Gene Transcripts

Gene Symbol
Robo2
Entrez Gene ID
268902
Gene Name
roundabout homolog 2 (Drosophila)
Gene Aliases
2600013A04Rik, 9430089E08Rik, BB097918, D230004I22Rik, mKIAA1568
Chromosome Location
Chr. 16: 73892061 - 75447298 on Build GRCm38
UniGene ID
Mm.171736
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175549.4 NP_780758.3 2 244
GenBank mRNA AK031495.1 - 2 770
AK035110.1 - 2 1118
AK048081.1 - 2 1158
AK051227.1 - 2 1158
AK085461.1 - 2 869
AK164874.1 - 2 868
BC055333.1 - 2 552 6409283
DQ533875.1 - 3 524
DQ533876.1 - 2 244

View Full Product Details