Gene Transcripts

Gene Symbol
Ptpn7
Entrez Gene ID
320139
Gene Name
protein tyrosine phosphatase, non-receptor type 7
Gene Aliases
BPTP-4, C920001D21Rik, HEPTP, LC-PTP, LPTP
Chromosome Location
Chr. 1: 135131840 - 135145328 on Build GRCm38
UniGene ID
Mm.258388
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177081.3 NP_796055.1 3, 4 420
GenBank mRNA AK041415.1 - 1, 2 1313
AK049175.1 - 2, 3 271
AK083305.1 - 3, 4 420
BC098475.1 - 3, 4 474 30044369
BC138194.1 - 2, 3 356 8861213
BC145190.1 - 2, 3 356 9053741

View Full Product Details