Gene Transcripts

Gene Symbol
Fbxl13
Entrez Gene ID
320118
Gene Name
F-box and leucine-rich repeat protein 13
Gene Aliases
4921539K22Rik
Chromosome Location
Chr. 5: 21483847 - 21645698 on Build GRCm38
UniGene ID
Mm.442295
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199632.1 NP_001186561.1 14, 15 1554
NM_177076.4 NP_796050.2 13, 14 1455
GenBank mRNA AK029566.1 - 13, 14 1457
AK030172.1 - 14, 15 1537
BC145291.1 - 14, 15 1480 9053845
BC145292.1 - 13, 14 1382 9053846

View Full Product Details