Gene Transcripts

Gene Symbol
Hykk
Entrez Gene ID
235386
Gene Name
hydroxylysine kinase 1
Gene Aliases
Agphd1, C630028N24Rik
Chromosome Location
Chr. 9: 54917290 - 54949924 on Build GRCm38
UniGene ID
Mm.259334
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_177351.4 NP_796325.2 2 745
GenBank mRNA AK048771.1 - 2 718
AK083218.1 - 1 3339
AK085527.1 - 2 754
AK138851.1 - 2 731
AK142509.1 - 1 3341
AK162425.1 - 2 746
AK163926.1 - 2 725
BC038618.1 - 2 724 4221933

View Full Product Details