Gene Transcripts

Gene Symbol
STYXL1
Entrez Gene ID
51657
Gene Name
serine/threonine/tyrosine interacting-like 1
Gene Aliases
DUSP24, MK-STYX, MKSTYX
Chromosome Location
Chr. 7: 75995820 - 76048709 on Build GRCh38
UniGene ID
Hs.11615
Species
Homo sapiens
Interrogated Sequence Translated Protein RNADB ID Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317785.1 NP_001304714.1
5, 6 807
NM_001317786.1 NP_001304715.1
5, 6 657
NM_016086.2 NP_057170.1
5, 6 804
NR_134487.1
4, 5 462
XM_011516292.2 XP_011514594.1
5, 6 650
XM_011516293.2 XP_011514595.1
5, 6 650
XM_017012296.1 XP_016867785.1
6, 7 764
XM_017012297.1 XP_016867786.1
5, 6 653
XM_017012298.1 XP_016867787.1
5, 6 613
XM_017012299.1 XP_016867788.1
6, 7 764
XM_017012300.1 XP_016867789.1
6, 7 684
GenBank mRNA AF069762.1 -
5, 6 800
AF169647.1 -
1 18
AF169648.1 -
1 18
AF188201.1 -
3, 4 326
AK308912.1 -
5, 6 631
AL533027.3 -
5, 6 605
AY927581.1 - LIT3580
AY927581
4, 5 443
BC024035.2 -
5, 6 767 2988900

View Full Product Details