Gene Transcripts

Gene Symbol
Ano1
Entrez Gene ID
101772
Gene Name
anoctamin 1, calcium activated chloride channel
Gene Aliases
Tmem16a
Chromosome Location
Chr. 7: 144588549 - 144752039 on Build GRCm38
UniGene ID
Mm.26700
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_178642.5 NP_848757.4 13, 15 1617
GenBank mRNA AK032793.1 - 8, 10 629
AK052589.1 - 13, 15 1569
BC004697.1 - 5, 7 483

View Full Product Details