Species:
Rat, Columba livia, Dasypus novemcinctus, Guinea pig, Ophiophagus hannah, Pteropus alecto, Python bivittatus, Rabbit, Taeniopygia guttata, Zebrafish
Mature miRNA Sequence:
GUCAUUUUUGUGAUGUUGCAGCU
Mapped to miRBase Version:
v22.1
miRBase Alias:
rno-miR-153* (v18)

Stem-loop Details

Stem-loop ID Stem-loop Accession # Mature miRNA ID Stem-loop Sequence Chromosome Location
rno-mir-153 MI0000922 rno-miR-153-5p AGCGGUGGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACUGUG Chr. 6 - 145105064 - 145105150 [+]
on Build Rnor_6.0
cli-mir-153b MI0029987 cli-miR-153b-5p UUAGCGGUUGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACUGUGAC -
cpo-mir-153 MI0038662 cpo-miR-153-5p GUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCA -
dno-mir-153b MI0039003 dno-miR-153b-5p GUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCA -
dre-mir-153a MI0002021 dre-miR-153a-5p GGUUGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUUAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACU -
ocu-mir-153 MI0039312 ocu-miR-153-5p GUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCA -
oha-mir-153-1 MI0031364 oha-miR-153-1-5p UGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUAAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAA -
pal-mir-153-1 MI0032475 pal-miR-153-5p GUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCA -
pbv-mir-153-1 MI0030250 pbv-miR-153-5p UAGCAGUUGCCAGUGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUAAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACUGU -
tgu-mir-153-1 MI0013768 tgu-miR-153-1-5p UGUCAUUUUUGUGAUGUUGCAGCUAGUAAUAUGAGCCCAGUUGCAUAGUCACAAAAGUGAUCAUUGGAAACU -

View Full Stem-loop Details