Species:
Mouse
Mature miRNA Sequence:
UUCACUGGAGUUUGUUUCAGU
Mapped to miRBase Version:
v22.1
miRBase Alias:
mmu-miR-653* (v17)

Stem-loop Details

Stem-loop ID Stem-loop Accession # Mature miRNA ID Stem-loop Sequence Chromosome Location
mmu-mir-653 MI0005557 mmu-miR-653-3p CAUUCUUUCAGUGUUGAAACAAUCUCUACUGAACCAAGCUCCAAAGCGAGUUCACUGGAGUUUGUUUCAGUAUUGCAGGAGUGCU Chr. 6 - 3721301 - 3721385 [-]
on Build GRCm38

View Full Stem-loop Details