Gene Transcripts

Gene Symbol
ACACB
Entrez Gene ID
32
Gene Name
acetyl-CoA carboxylase beta
Gene Aliases
ACC2, ACCB, HACC275
Chromosome Location
Chr. 12: 109112666 - 109268226 on Build GRCh38
UniGene ID
Hs.234898
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001093.3 NP_001084.3 39 5621
XM_005253876.4 XP_005253933.1 40 5731
XM_006719367.3 XP_006719430.1 39 5252
XM_011538259.2 XP_011536561.1 40 6369
XM_011538263.2 XP_011536565.1 39 6180
XM_011538264.2 XP_011536566.1 39 5706
XM_017019252.1 XP_016874741.1 38 5047
GenBank mRNA AB209110.1 - 29 3675
AJ575592.3 - 39 5612
AK302469.1 - 12 1550
AK302502.1 - 13 1683
AY382667.1 - 40 5731
BC028417.1 - 13 1707
DQ493870.1 - 39 5612
U34591.1 - 13 1643
U89344.1 - 38 5687

View Full Product Details