Gene Transcripts

Gene Symbol
C1S
Entrez Gene ID
716
Gene Name
complement component 1, s subcomponent
Gene Aliases
Chromosome Location
Chr. 12: 7060676 - 7071032 on Build GRCh38
UniGene ID
Hs.458355
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001734.3 NP_001725.1 12 1720
NM_201442.2 NP_958850.1 12 1807
XM_005253760.1 XP_005253817.1 12 1699
GenBank mRNA AK025309.1 - 9 1195
AK290295.1 - 12 1754
AM886411.1 - 11 1442
BC056903.1 - 12 1747 6199186
J04080.1 - 12 1646
M18767.1 - 12 1666

View Full Product Details