Gene Transcripts

Gene Symbol
CTSB
Entrez Gene ID
1508
Gene Name
cathepsin B
Gene Aliases
APPS, CPSB
Chromosome Location
Chr. 8: 11842524 - 11868150 on Build GRCh38
UniGene ID
Hs.520898
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317237.1 NP_001304166.1 3 348
NM_001908.4 NP_001899.1 2 260
NM_147780.3 NP_680090.1 4 422
NM_147781.3 NP_680091.1 3 379
NM_147782.3 NP_680092.1 3 348
NM_147783.3 NP_680093.1 3 334
XM_006716244.2 XP_006716307.1 4 446
XM_006716245.2 XP_006716308.1 2 550
XM_011543812.2 XP_011542114.1 4 355
XM_017013097.1 XP_016868586.1 3 336
XM_017013098.1 XP_016868587.1 3 336
XM_017013099.1 XP_016868588.1 4 378
XM_017013100.1 XP_016868589.1 3 280
GenBank mRNA AK075393.1 - 2 236
AK130184.1 - 2 197
AK290239.1 - 2 197
AL543654.3 - 3 308
BC095408.1 - 2 197 30334082
BG770805.1 - 4 331
BX648239.1 - 1 1031
L16510.1 - 3 262

View Full Product Details