Gene Transcripts

Gene Symbol
EIF2S3
Entrez Gene ID
1968
Gene Name
eukaryotic translation initiation factor 2 subunit gamma
Gene Aliases
EIF2, EIF2G, EIF2gamma, eIF-2gA
Chromosome Location
Chr. X: 24054948 - 24078810 on Build GRCh38
UniGene ID
Hs.539684
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001415.3 NP_001406.1 4 319
GenBank mRNA AB451230.1 - 4 298
AB451353.1 - 4 298
AK222577.1 - 4 311
AK290397.1 - 4 311
AK312247.1 - 4 311
BC019906.1 - 4 319 4419438
L19161.1 - 4 298

View Full Product Details