Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000804.3 | NP_000795.2 | 5 | 787 | |
NM_001318045.1 | NP_001304974.1 | 5 | 915 | ||
XM_011544876.1 | XP_011543178.1 | 3 | 451 | ||
GenBank mRNA | CD368314.1 | - | 1 | 92 | |
U08470.1 | - | 4 | 731 | ||
U08471.1 | - | 4 | 733 | ||
Z32564.1 | - | 4 | 733 | ||
Z32633.1 | - | 4 | 731 |