Gene Transcripts

Gene Symbol
GDF10
Entrez Gene ID
2662
Gene Name
growth differentiation factor 10
Gene Aliases
BMP-3b, BMP3B
Chromosome Location
Chr. 10: 47300224 - 47313577 on Build GRCh38
UniGene ID
Hs.2171
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004962.3 NP_004953.1 3 1802
GenBank mRNA AK315322.1 - 3 1655
BC028237.1 - 3 1232 5245100
D49492.1 - 3 1640
L42113.1 - 2 297

View Full Product Details