Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166425.1 | NP_001159897.1 | 8 | 1087 | |
NM_001282617.1 | NP_001269546.1 | 8 | 1128 | ||
NM_001282618.1 | NP_001269547.1 | 7 | 1159 | ||
NM_001282619.1 | NP_001269548.1 | 9 | 1535 | ||
NM_001282620.1 | NP_001269549.1 | 8 | 1411 | ||
NM_002070.3 | NP_002061.1 | 8 | 1202 | ||
GenBank mRNA | AF493906.1 | - | 8 | 1023 | |
AK055574.1 | - | 7 | 1112 | ||
AK096299.1 | - | 8 | 1304 | ||
AK096595.1 | - | 8 | 1806 | ||
AK126708.1 | - | 8 | 2839 | ||
AK131062.1 | - | 6 | 1850 | ||
AK302330.1 | - | 8 | 1056 | ||
AK304473.1 | - | 8 | 967 | ||
BC012138.1 | - | 8 | 1183 | 4562585 | |
BC014627.2 | - | 8 | 1140 | 2963802 | |
BC016995.1 | - | 9 | 1486 | 4403631 | |
CR456783.1 | - | 8 | 1023 | ||
X04828.1 | - | 8 | 1146 |