Gene Transcripts

Gene Symbol
IL1A
Entrez Gene ID
3552
Gene Name
interleukin 1 alpha
Gene Aliases
IL-1A, IL1, IL1-ALPHA, IL1F1
Chromosome Location
Chr. 2: 112773915 - 112785398 on Build GRCh38
UniGene ID
Hs.1722
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000575.4 NP_000566.3 6 1478
GenBank mRNA AK314850.1 - 6 573
BC013142.2 - 5 523 3875617
BT007014.1 - 5 514
CR457414.1 - 5 514
M15329.1 - 5 519
X02531.1 - 5 514
X02851.1 - 6 572
X56086.1 - 6 540

View Full Product Details