Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000577.4 | NP_000568.1 | 3 | 205 | |
NM_001318914.1 | NP_001305843.1 | 5 | 439 | ||
NM_173841.2 | NP_776213.1 | 4 | 268 | ||
NM_173842.2 | NP_776214.1 | 2 | 197 | ||
NM_173843.2 | NP_776215.1 | 4 | 376 | ||
XM_005263661.4 | XP_005263718.1 | 3 | 224 | ||
XM_011511121.1 | XP_011509423.1 | 7 | 727 | ||
GenBank mRNA | AF043143.1 | - | 4 | 376 | |
AK290898.1 | - | 4 | 258 | ||
AK290926.1 | - | 2 | 199 | ||
BC009745.2 | - | 3 | 110 | 3689284 | |
BI766516.1 | - | 2 | 197 | ||
DA667440.1 | - | 5 | 356 | ||
M55646.1 | - | 3 | 201 | ||
X52015.1 | - | 2 | 156 | ||
X53296.1 | - | 2 | 141 | ||
X84348.1 | - | 4 | 174 |