Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005546.3 | NP_005537.3 | 3, 4 | 405 | |
GenBank mRNA | AB209622.1 | - | 3, 4 | 458 | |
AK303609.1 | - | 3, 4 | 371 | ||
AK312846.1 | - | 3, 4 | 374 | ||
BC109077.2 | - | 3, 4 | 396 | 40034274 | |
BC109078.1 | - | 3, 4 | 396 | 40034276 | |
D13720.1 | - | 3, 4 | 405 |