Gene Transcripts

Gene Symbol
MARK1
Entrez Gene ID
4139
Gene Name
microtubule affinity regulating kinase 1
Gene Aliases
MARK, Par-1c, Par1c
Chromosome Location
Chr. 1: 220528183 - 220664457 on Build GRCh38
UniGene ID
Hs.497806
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001286124.1 NP_001273053.1 16 2513
NM_001286126.1 NP_001273055.1 16 2510
NM_001286128.1 NP_001273057.1 15 2444
NM_018650.4 NP_061120.3 16 2510
XM_005273134.4 XP_005273191.1 16 2380
XM_006711326.3 XP_006711389.1 16 1774
XM_011509561.2 XP_011507863.1 16 2509
XM_011509562.2 XP_011507864.1 16 2126
XM_017001305.1 XP_016856794.1 17 1970
XM_017001306.1 XP_016856795.1 16 2007
XM_017001307.1 XP_016856796.1 17 2407
GenBank mRNA AB040910.1 - 15 2401
AF154845.1 - 16 2136
AK027493.1 - 12 1467
AK295506.1 - 16 2008
AK314614.1 - 16 2173
BC113869.1 - 16 2046 40036731
BC114478.1 - 16 2046 40036730
KJ534879.1 - 16 1873
KJ534880.1 - 10 1270
KJ534881.1 - 16 1870
KJ535089.1 - 16 1869

View Full Product Details