Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006739.3 | NP_006730.2 | 8 | 1083 | |
XM_006724242.3 | XP_006724305.1 | 8 | 1138 | ||
GenBank mRNA | AB209612.1 | - | 2 | 571 | |
AK122853.1 | - | 7 | 931 | ||
AK130620.1 | - | 7 | 1037 | ||
AK223323.1 | - | 8 | 1073 | ||
AK291136.1 | - | 8 | 1064 | ||
AK298624.1 | - | 8 | 960 | ||
AK301121.1 | - | 1225 | |||
AK308343.1 | - | 7 | 796 | ||
AK312533.1 | - | 8 | 1063 | ||
BC000142.1 | - | 8 | 1035 | 2900229 | |
BC003656.2 | - | 7 | 1009 | 3542201 | |
CR456517.1 | - | 8 | 1023 | ||
D83986.1 | - | 8 | 1071 | ||
X74795.2 | - | 7 | 1011 |