Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_005937.3 | NP_005928.2 | 18, 19 | 2907 | |
XM_006721911.2 | XP_006721974.1 | 18, 19 | 2829 | ||
XM_017024648.1 | XP_016880137.1 | 13, 14 | 2432 | ||
GenBank mRNA | AB209633.1 | - | 18, 19 | 2882 | |
AK027133.1 | - | 7, 8 | 1025 | ||
AL133659.1 | - | 6, 7 | 1886 | ||
BC007237.1 | - | 10, 11 | 1858 | ||
BX648298.1 | - | 1 | 375 | ||
U07932.1 | - | 18, 19 | 2894 |