Gene Transcripts

Gene Symbol
MUC4
Entrez Gene ID
4585
Gene Name
mucin 4, cell surface associated
Gene Aliases
ASGP, HSA276359, MUC-4
Chromosome Location
Chr. 3: 195746765 - 195811973 on Build GRCh38
UniGene ID
Hs.369646
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001322468.1 NP_001309397.1 19263
NM_004532.5 NP_004523.3 4 537
NM_018406.6 NP_060876.5 13245
NM_138297.4 NP_612154.2 3 384
GenBank mRNA AJ010901.1 - 4 321
AJ242541.1 - 4 335
AJ242542.1 - 4 335
AJ242546.1 - 4 321
AJ242547.1 - 4 321
AJ242548.1 - 4 321
AJ242549.1 - 4 421
AJ242550.1 - 3 268
AJ276359.1 - 6 3431
AJ277412.1 - 6 3431
AJ277505.1 - 6 3417
AJ400633.1 - 6 3445
AJ400851.1 - 6 3445
AJ400854.1 - 6 3431
AJ400855.1 - 6 3431
AJ400856.1 - 6 3431
AJ400857.1 - 6 3431
AJ400858.1 - 6 3431
BX957139.1 - 3 292
DB227644.1 - 2 131
EF091824.1 - 7 3306

View Full Product Details