Gene Transcripts

Gene Symbol
NEO1
Entrez Gene ID
4756
Gene Name
neogenin 1
Gene Aliases
IGDCC2, NGN, NTN1R2
Chromosome Location
Chr. 15: 73051715 - 73305206 on Build GRCh38
UniGene ID
Hs.388613
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172623.1 NP_001166094.1 24 3666
NM_001172624.1 NP_001166095.1 23 3633
NM_002499.3 NP_002490.2 24 3666
XM_005254408.1 XP_005254465.1 25 3976
XM_011521628.2 XP_011519930.1 24 3731
XM_011521629.2 XP_011519931.1 23 3692
XM_011521630.2 XP_011519932.1 24 3677
XM_011521632.2 XP_011519934.1 22 3623
XM_011521633.2 XP_011519935.1 24 3725
XM_017022231.1 XP_016877720.1 23 3584
XM_017022232.1 XP_016877721.1 24 3895
XM_017022233.1 XP_016877722.1 23 3692
XM_017022234.1 XP_016877723.1 25 3720
XM_017022235.1 XP_016877724.1 23 3644
XM_017022236.1 XP_016877725.1 23 3632
XM_017022237.1 XP_016877726.1 22 3623
XM_017022238.1 XP_016877727.1 23 3584
GenBank mRNA AB209412.1 - 19 2481
BC117161.1 - 24 3540 40125712
BC143270.1 - 24 3541 9051772
BC143271.1 - 23 3507 9051773
U61262.1 - 24 3610
U72391.1 - 24 3600

View Full Product Details