Gene Transcripts

Gene Symbol
NOTCH2
Entrez Gene ID
4853
Gene Name
notch 2
Gene Aliases
AGS2, HJCYS, hN2
Chromosome Location
Chr. 1: 119911553 - 120073449 on Build GRCh38
UniGene ID
Hs.487360
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001200001.1 NP_001186930.1 20 3502
NM_024408.3 NP_077719.2 20 3502
XM_005270901.3 XP_005270958.1 21 5816
XM_011541519.2 XP_011539821.1 20 8610
XM_011541520.2 XP_011539822.1 20 5520
XM_017001372.1 XP_016856861.1 20 3172
XM_017001373.1 XP_016856862.1 20 3151
GenBank mRNA AF308601.1 - 20 3217
AF315356.1 - 20 3217
BC071562.1 - 20 3451 4822955
U77493.1 - 3 307

View Full Product Details