Gene Transcripts

Gene Symbol
OTX2
Entrez Gene ID
5015
Gene Name
orthodenticle homeobox 2
Gene Aliases
CPHD6, MCOPS5
Chromosome Location
Chr. 14: 56800707 - 56810476 on Build GRCh38
UniGene ID
Hs.288655
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001270523.1 NP_001257452.1 5 886
NM_001270524.1 NP_001257453.1 4 1039
NM_001270525.1 NP_001257454.1 3 833
NM_021728.3 NP_068374.1 5 910
NM_172337.2 NP_758840.1 3 809
NR_073034.1 4 734
NR_073036.1 2 657
GenBank mRNA AB593056.1 - 2 660
AB593057.1 - 3 579
AB593058.1 - 5 910
AF093138.1 - 3 602
AK314271.1 - 4 942
BC032579.1 - 5 911 5493541

View Full Product Details