Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_002567.3 | NP_002558.1 | 4 | 849 | |
GenBank mRNA | AK226006.1 | - | 4 | 628 | |
AK308056.1 | - | 4 | 431 | ||
AK311927.1 | - | 4 | 620 | ||
BC008714.2 | - | 4 | 622 | 2822606 | |
BC017396.1 | - | 4 | 643 | 4074533 | |
BC018799.1 | - | 4 | 613 | ||
BC031102.1 | - | 4 | 611 | 5300366 | |
D16111.1 | - | 4 | 614 | ||
EU668351.1 | - | 3 | 382 | ||
EU794643.1 | - | 4 | 613 | ||
GQ472218.1 | - | 3 | 382 | ||
HM131449.1 | - | 4 | 605 | ||
S76773.1 | - | 3 | 382 | ||
X75252.1 | - | 4 | 633 | ||
X85033.1 | - | 4 | 526 |