Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001316329.1 | NP_001303258.1 | 5 | 823 | |
NM_002739.4 | NP_002730.1 | 5 | 823 | ||
GenBank mRNA | AB209671.1 | - | 5 | 542 | |
AF345987.1 | - | 5 | 707 | ||
AK296412.1 | - | 5 | 831 | ||
AK296434.1 | - | 5 | 810 | ||
AK302958.1 | - | 5 | 808 | ||
AK303741.1 | - | 3 | 230 | ||
AK312302.1 | - | 5 | 785 | ||
BC047876.1 | - | 5 | 698 | 5764695 | |
M13977.1 | - | 5 | 707 | ||
Z15114.1 | - | 1 | 31 |