Gene Transcripts

Gene Symbol
PRKCQ
Entrez Gene ID
5588
Gene Name
protein kinase C theta
Gene Aliases
PRKCT, nPKC-theta
Chromosome Location
Chr. 10: 6393038 - 6580646 on Build GRCh38
UniGene ID
Hs.498570
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001242413.2 NP_001229342.1 16, 17 1874
NM_001282644.1 NP_001269573.1 17, 18 2061
NM_001282645.1 NP_001269574.1 16, 17 2002
NM_001323265.1 NP_001310194.1 17, 18 2118
NM_001323266.1 NP_001310195.1 16, 17 2000
NM_006257.4 NP_006248.1 17, 18 2063
XM_005252496.4 XP_005252553.1 17, 18 2163
XM_006717465.3 XP_006717528.1 19, 20 4558
XM_017016410.1 XP_016871899.1 18, 19 4469
XM_017016411.1 XP_016871900.1 18, 19 4497
GenBank mRNA AK293935.1 - 16, 17 1940
AK308099.1 - 17, 18 2036
AK312411.1 - 17, 18 2014
AY702977.1 - 15, 16 1774
BC101465.1 - 16, 17 1982 8068971
BC113359.1 - 16, 17 1982 8322411
BX647657.1 - 16, 17 2148
L01087.1 - 17, 18 2057
L07032.1 - 16, 17 1984

View Full Product Details