Gene Transcripts

Gene Symbol
NECTIN2
Entrez Gene ID
5819
Gene Name
nectin cell adhesion molecule 2
Gene Aliases
CD112, HVEB, PRR2, PVRL2, PVRR2
Chromosome Location
Chr. 19: 44846136 - 44889228 on Build GRCh38
UniGene ID
Hs.655455
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001042724.1 NP_001036189.1 5 1375
NM_002856.2 NP_002847.1 5 1375
XM_011527192.2 XP_011525494.1 5 1375
GenBank mRNA AF058448.1 - 5 1134
AK291330.1 - 5 1201
BC003091.1 - 5 1183 3503222
CR456818.1 - 5 985
X80038.1 - 5 985

View Full Product Details