Gene Transcripts

Gene Symbol
RAD9A
Entrez Gene ID
5883
Gene Name
RAD9 checkpoint clamp component A
Gene Aliases
RAD9
Chromosome Location
Chr. 11: 67391952 - 67398412 on Build GRCh38
UniGene ID
Hs.655354
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001243224.1 NP_001230153.1 5 652
NM_004584.2 NP_004575.1 8 788
XM_006718652.3 XP_006718715.1 6 1000
XM_017018097.1 XP_016873586.1 8 732
XM_017018098.1 XP_016873587.1 6 524
GenBank mRNA AK310213.1 - 4 1164
AK315348.1 - 8 727
BC014848.1 - 8 788 4508954
CR536508.1 - 8 695
U53174.1 - 8 771

View Full Product Details