Gene Transcripts

Gene Symbol
SLC5A4
Entrez Gene ID
6527
Gene Name
solute carrier family 5 member 4
Gene Aliases
DJ90G24.4, SAAT1, SGLT3
Chromosome Location
Chr. 22: 32218423 - 32355057 on Build GRCh38
UniGene ID
Hs.130101
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014227.2 NP_055042.1 1 61
GenBank mRNA AJ133127.1 - 1 75

View Full Product Details