Gene Transcripts

Gene Symbol
SPTAN1
Entrez Gene ID
6709
Gene Name
spectrin alpha, non-erythrocytic 1
Gene Aliases
EIEE5, NEAS, SPTA2
Chromosome Location
Chr. 9: 128552558 - 128633665 on Build GRCh38
UniGene ID
Hs.372331
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001130438.2 NP_001123910.1 44 5777
NM_001195532.1 NP_001182461.1 42 5702
NM_003127.3 NP_003118.2 43 5762
XM_006717245.1 XP_006717308.1 44 5684
XM_006717246.1 XP_006717309.1 43 5669
XM_006717247.2 XP_006717310.1 43 5624
XM_006717248.2 XP_006717311.1 44 5683
XM_006717249.1 XP_006717312.1 43 5669
XM_006717250.1 XP_006717313.1 44 5684
XM_006717251.2 XP_006717314.1 43 5711
XM_006717252.2 XP_006717315.1 43 5624
XM_006717253.1 XP_006717316.1 42 5609
XM_006717254.2 XP_006717317.1 44 5771
XM_017015059.1 XP_016870548.1 44 5762
XM_017015060.1 XP_016870549.1 43 5624
XM_017015061.1 XP_016870550.1 43 5682
GenBank mRNA AB191262.1 - 42 5667
AB209860.1 - 43 5724
AL110273.1 - 22 2589
BC053521.1 - 42 5651 5529391
J05243.1 - 43 5722
M19725.1 - 4 311
U83867.1 - 44 5704

View Full Product Details