Gene Transcripts

Gene Symbol
SPTBN2
Entrez Gene ID
6712
Gene Name
spectrin beta, non-erythrocytic 2
Gene Aliases
GTRAP41, SCA5, SCAR14
Chromosome Location
Chr. 11: 66682496 - 66729361 on Build GRCh38
UniGene ID
Hs.26915
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006946.2 NP_008877.1 20 4326
XM_005274192.4 XP_005274249.1 21 4988
XM_005274193.3 XP_005274250.1 20 4762
XM_006718669.3 XP_006718732.1 19 6675
XM_006718671.3 XP_006718734.1 21 4388
XM_017018174.1 XP_016873663.1 21 5080
XM_017018175.1 XP_016873664.1 21 4709
XM_017018176.1 XP_016873665.1 21 4429
XM_017018177.1 XP_016873666.1 20 4618
XM_017018178.1 XP_016873667.1 20 4301
GenBank mRNA AB008567.1 - 20 4326
BC131819.1 - 4 588

View Full Product Details