Gene Transcripts

Gene Symbol
SSR2
Entrez Gene ID
6746
Gene Name
signal sequence receptor subunit 2
Gene Aliases
HSD25, TLAP, TRAP-BETA, TRAPB
Chromosome Location
Chr. 1: 156009048 - 156020992 on Build GRCh38
UniGene ID
Hs.74564
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003145.3 NP_003136.1 2 200
XM_011509923.2 XP_011508225.1 2 225
GenBank mRNA AK222600.1 - 2 143
AK300678.1 - 2 200
AK313821.1 - 2 200
AY251536.1 - 2 160
BC000341.2 - 2 146 2822983
BP196192.1 - 2 200
BX649192.1 - 2 185
CR456973.1 - 1 120
D37991.1 - 2 144
X74104.1 - 2 170

View Full Product Details