Gene Transcripts

Gene Symbol
TMOD1
Entrez Gene ID
7111
Gene Name
tropomodulin 1
Gene Aliases
D9S57E, ETMOD, TMOD
Chromosome Location
Chr. 9: 97501180 - 97601743 on Build GRCh38
UniGene ID
Hs.404289
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001166116.1 NP_001159588.1 8 896
NM_003275.3 NP_003266.1 8 973
GenBank mRNA AK095748.1 - 5 640
AK314533.1 - 8 896
BC002660.2 - 8 973 3610014
M77016.1 - 7 794

View Full Product Details