Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003365.2 | NP_003356.2 | 8 | 882 | |
GenBank mRNA | AK129903.1 | - | 8 | 862 | |
AK300694.1 | - | 8 | 817 | ||
AK313090.1 | - | 8 | 852 | ||
BC009586.1 | - | 8 | 853 | 4096628 | |
D26485.1 | - | 8 | 844 | ||
L16842.1 | - | 8 | 1254 |