Gene Transcripts

Gene Symbol
USP1
Entrez Gene ID
7398
Gene Name
ubiquitin specific peptidase 1
Gene Aliases
UBP
Chromosome Location
Chr. 1: 62436304 - 62451804 on Build GRCh38
UniGene ID
Hs.35086
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001017415.1 NP_001017415.1 8, 9 1944
NM_001017416.1 NP_001017416.1 8, 9 1849
NM_003368.4 NP_003359.3 8, 9 2374
GenBank mRNA AB014458.1 - 8, 9 1725
AB208893.1 - 8, 9 2005
AF117386.1 - 8, 9 1861
AK312423.1 - 8, 9 2037
AL117575.1 - 7, 8 1696
BC023216.1 - 4, 5 1209
BC050525.1 - 8, 9 2078 6473568

View Full Product Details