Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007124.2 | NP_009055.2 | 22 | 2970 | |
XM_005267127.4 | XP_005267184.1 | 22 | 3305 | ||
XM_005267130.2 | XP_005267187.1 | 23 | 3188 | ||
XM_005267133.2 | XP_005267190.1 | 22 | 3988 | ||
XM_011536101.2 | XP_011534403.1 | 24 | 3649 | ||
XM_011536102.2 | XP_011534404.1 | 24 | 3190 | ||
XM_011536106.2 | XP_011534408.1 | 23 | 3837 | ||
XM_011536107.2 | XP_011534409.1 | 23 | 3213 | ||
XM_017011243.1 | XP_016866732.1 | 24 | 3187 | ||
XM_017011244.1 | XP_016866733.1 | 24 | 3190 | ||
XM_017011245.1 | XP_016866734.1 | 24 | 3190 | ||
GenBank mRNA | AK292506.1 | - | 23 | 3185 | |
X69086.1 | - | 22 | 2878 |