Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004625.3 | NP_004616.2 | 3 | 845 | |
XM_011534091.2 | XP_011532393.1 | 4 | 592 | ||
GenBank mRNA | BC008811.2 | - | 3 | 845 | |
D83175.1 | - | 3 | 570 | ||
U53476.1 | - | 3 | 564 |