Gene Transcripts

Gene Symbol
CUL1
Entrez Gene ID
8454
Gene Name
cullin 1
Gene Aliases
Chromosome Location
Chr. 7: 148698178 - 148801110 on Build GRCh38
UniGene ID
Hs.146806
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_003592.2 NP_003583.2 3 598
XM_005250060.4 XP_005250117.1 3 634
XM_011516629.2 XP_011514931.1 3 590
XM_011516630.2 XP_011514932.1 3 491
XM_011516631.2 XP_011514933.1 3 619
XM_011516632.2 XP_011514934.1 3 517
XM_017012723.1 XP_016868212.1 3 518
GenBank mRNA AF062536.1 - 2 372
AK096163.1 - 4 486
AK315023.1 - 3 774
BC125119.1 - 3 511 40118923
BC125120.1 - 3 511 40118927
BG391038.1 - 613
BX537409.1 - 3 528

View Full Product Details