Gene Transcripts
- Gene Symbol
- CUL1
- Entrez Gene ID
- 8454
- Gene Name
- cullin 1
- Gene Aliases
- Chromosome Location
- Chr. 7: 148698178 - 148801110 on Build GRCh38
- UniGene ID
- Hs.146806
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_003592.2 |
NP_003583.2
|
5, 6 |
878 |
|
XM_005250060.4 |
XP_005250117.1
|
5, 6 |
914 |
|
XM_011516629.2 |
XP_011514931.1
|
5, 6 |
870 |
|
XM_011516630.2 |
XP_011514932.1
|
5, 6 |
771 |
|
XM_011516631.2 |
XP_011514933.1
|
5, 6 |
899 |
|
XM_011516632.2 |
XP_011514934.1
|
5, 6 |
797 |
|
XM_017012723.1 |
XP_016868212.1
|
5, 6 |
798 |
|
GenBank mRNA |
AF062536.1 |
- |
4, 5 |
652 |
|
AK096163.1 |
- |
6, 7 |
766 |
|
AK296909.1 |
- |
4, 5 |
603 |
|
AK315023.1 |
- |
5, 6 |
1054 |
|
BC125119.1 |
- |
5, 6 |
791 |
40118923 |
BC125120.1 |
- |
5, 6 |
791 |
40118927 |
BX537409.1 |
- |
5, 6 |
808 |
|
U58087.1 |
- |
5, 6 |
580 |
|
View Full Product Details