Gene Transcripts

Gene Symbol
GBF1
Entrez Gene ID
8729
Gene Name
golgi brefeldin A resistant guanine nucleotide exchange factor 1
Gene Aliases
ARF1GEF
Chromosome Location
Chr. 10: 102245498 - 102382899 on Build GRCh38
UniGene ID
Hs.290243
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001199378.1 NP_001186307.1 20 2815
NM_001199379.1 NP_001186308.1 20 2812
NM_004193.2 NP_004184.1 20 2812
XM_005270261.1 XP_005270318.1 20 2815
XM_006718047.1 XP_006718110.1 21 2914
XM_006718048.1 XP_006718111.1 21 2911
XM_006718049.1 XP_006718112.1 21 2914
XM_006718050.1 XP_006718113.1 21 2878
XM_011540312.1 XP_011538614.1 20 2773
XM_011540313.1 XP_011538615.1 21 2914
XM_011540314.2 XP_011538616.1 9 1292
XM_017016861.1 XP_016872350.1 21 2890
XM_017016862.1 XP_016872351.1 21 2878
XM_017016863.1 XP_016872352.1 20 2776
XM_017016864.1 XP_016872353.1 16 2114
GenBank mRNA AF068755.1 - 20 2758
AK025330.1 - 20 2781
BC117681.1 - 20 2639 40070852
BC117682.1 - 20 2639 40070853
D87435.2 - 20 2742

View Full Product Details