Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_006773.3 | NP_006764.3 | 9 | 1429 | |
GenBank mRNA | AB209392.1 | - | 2 | 496 | |
AK001467.1 | - | 9 | 1401 | ||
AK091227.1 | - | 7 | 856 | ||
BC001238.1 | - | 9 | 1358 | 3452935 | |
BC003360.1 | - | 9 | 1388 | 2900252 | |
BC024739.1 | - | 9 | 1403 | 4995871 | |
CR457060.1 | - | 8 | 1150 | ||
X98743.1 | - | 8 | 1167 |