Gene Transcripts

Gene Symbol
DYRK1B
Entrez Gene ID
9149
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 1B
Gene Aliases
AOMS3, MIRK
Chromosome Location
Chr. 19: 39825347 - 39834233 on Build GRCh38
UniGene ID
Hs.130988
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004714.2 NP_004705.1 4, 5 690
NM_006483.2 NP_006474.1 4, 5 690
NM_006484.2 NP_006475.1 4, 5 690
XM_005259398.4 XP_005259455.1 4, 5 1142
GenBank mRNA AF205861.1 - 4, 5 587
AK074971.1 - 4, 5 606
AK302127.1 - 4, 5 619
BC018751.1 - 4, 5 642 4300012
BC025291.1 - 4, 5 593 5019417
Y17999.1 - 4, 5 658

View Full Product Details