Gene Transcripts

Gene Symbol
NUMBL
Entrez Gene ID
9253
Gene Name
NUMB like, endocytic adaptor protein
Gene Aliases
CAG3A, CTG3a, NBL, NUMB-R, NUMBLIKE, NUMBR, TNRC23
Chromosome Location
Chr. 19: 40665905 - 40690658 on Build GRCh38
UniGene ID
Hs.326953
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001289979.1 NP_001276908.1 5 552
NM_001289980.1 NP_001276909.1 5 436
NM_004756.4 NP_004747.1 6 637
GenBank mRNA AF015041.1 - 6 671
AK289398.1 - 6 629
AK299484.1 - 5 436
BC001794.1 - 5 525
BC144583.1 - 5 447
BT009807.1 - 4 339

View Full Product Details