Gene Transcripts
- Gene Symbol
- SLC9A3R2
- Entrez Gene ID
- 9351
- Gene Name
- SLC9A3 regulator 2
- Gene Aliases
- E3KARP, NHE3RF2, NHERF-2, NHERF2, OCTS2, SIP-1, SIP1, TKA-1
- Chromosome Location
- Chr. 16: 2026868 - 2039026 on Build GRCh38
- UniGene ID
- Hs.440896
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001130012.2 |
NP_001123484.1
|
2, 3 |
545 |
|
NM_004785.5 |
NP_004776.3
|
2, 3 |
545 |
|
XM_017023895.1 |
XP_016879384.1
|
2, 3 |
400 |
|
GenBank mRNA |
AF004900.1 |
- |
2, 3 |
423 |
|
AF035771.1 |
- |
2, 3 |
407 |
|
BC014513.1 |
- |
2, 3 |
518 |
|
BC069014.1 |
- |
2, 3 |
514 |
|
BC106001.1 |
- |
2, 3 |
521 |
5296143 |
DB495688.2 |
- |
2, 3 |
526 |
|
U82108.1 |
- |
2, 3 |
498 |
|
Z50150.1 |
- |
2, 3 |
500 |
|
View Full Product Details