Gene Transcripts

Gene Symbol
UBE4A
Entrez Gene ID
9354
Gene Name
ubiquitination factor E4A
Gene Aliases
E4, UBOX2, UFD2
Chromosome Location
Chr. 11: 118359581 - 118399211 on Build GRCh38
UniGene ID
Hs.602967
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001204077.1 NP_001191006.1 9 1350
NM_004788.3 NP_004779.2 9 1371
GenBank mRNA AK296369.1 - 9 1297
AK307737.1 - 4 548
BC062695.1 - 9 1330
BC111417.1 - 9 1324 40038776
BC112367.1 - 9 1327 40038767
D50916.1 - 9 1308

View Full Product Details