Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004273.4 | NP_004264.2 | 2, 3 | 590 | |
XM_006718075.3 | XP_006718138.1 | 2, 3 | 823 | ||
XM_011540369.2 | XP_011538671.1 | 2, 3 | 2516 | ||
GenBank mRNA | AB012192.1 | - | 2, 3 | 584 | |
AB017915.1 | - | 2, 3 | 590 | ||
BC093690.1 | - | 1, 2 | 199 | 7939535 | |
BC104856.1 | - | 1, 2 | 199 | 8143859 |