Gene Transcripts

Gene Symbol
SLC4A7
Entrez Gene ID
9497
Gene Name
solute carrier family 4 member 7
Gene Aliases
NBC2, NBC3, NBCN1, SBC2, SLC4A6
Chromosome Location
Chr. 3: 27372721 - 27484420 on Build GRCh38
UniGene ID
Hs.250072
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001258379.1 NP_001245308.1 19 2841
NM_001258380.1 NP_001245309.1 19 2603
NM_001321103.1 NP_001308032.1 20 3225
NM_001321104.1 NP_001308033.1 20 3186
NM_001321105.1 NP_001308034.1 20 3186
NM_001321106.1 NP_001308035.1 20 3174
NM_001321107.1 NP_001308036.1 19 2841
NM_001321108.1 NP_001308037.1 19 2853
NM_003615.4 NP_003606.3 20 2975
NR_135541.1 19 2999
NR_135542.1 18 2666
NR_135543.1 18 2666
NR_135544.1 19 2999
XM_005265598.4 XP_005265655.1 19 3107
XM_005265600.4 XP_005265657.1 19 3069
XM_006713421.2 XP_006713484.1 20 3213
XM_011534256.2 XP_011532558.1 20 3486
XM_011534258.1 XP_011532560.1 20 3189
XM_011534261.2 XP_011532563.1 20 3451
XM_011534263.2 XP_011532565.1 19 2778
XM_011534265.1 XP_011532567.1 19 2817
XM_017007526.1 XP_016863015.1 21 3110
XM_017007527.1 XP_016863016.1 19 2802
XM_017007528.1 XP_016863017.1 13 1726
XM_017007529.1 XP_016863018.1 13 1708
GenBank mRNA AB012130.1 - 16 2289
AF047033.1 - 20 2975
AF053755.1 - 16 2281
AF089726.1 - 19 2603
CR627428.1 - 12 1557
EU499349.1 - 19 2768
EU934246.1 - 20 3101
EU934247.1 - 20 3113
EU934248.1 - 20 3152
EU934249.1 - 19 2768
EU934250.1 - 20 3113
FJ178574.1 - 19 2926
FJ178575.1 - 18 2593
FJ178576.1 - 18 2593
GU354307.1 - 19 2926
GU354308.1 - 19 2780
GU354309.1 - 18 2605
GU354310.1 - 18 2605

View Full Product Details