Gene Transcripts

Gene Symbol
TBKBP1
Entrez Gene ID
9755
Gene Name
TBK1 binding protein 1
Gene Aliases
ProSAPiP2, SINTBAD
Chromosome Location
Chr. 17: 47694065 - 47712063 on Build GRCh38
UniGene ID
Hs.94790
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014726.2 NP_055541.1 2, 3 1185
XM_005257859.4 XP_005257916.1 3, 4 484
XM_005257860.4 XP_005257917.1 3, 4 1309
XM_005257861.4 XP_005257918.1 3, 4 789
XM_005257862.3 XP_005257919.1 3, 4 409
XM_017025431.1 XP_016880920.1 2, 3 1693
GenBank mRNA AB018318.1 - 2, 3 1185
BC111418.1 - 2, 3 764
BC152407.1 - 2, 3 1185 8862510
BC167150.1 - 2, 3 1185 8862522

View Full Product Details